6-Fluoro-1H-benzo[d]imidazole-2-carbaldehyde 95+%

Supplier: Apollo Scientific

PC201355-100MG PC201355-250MG PC201355-1G
APOSPC201355-100MGEA 87.3 EUR
APOSPC201355-100MG APOSPC201355-250MG APOSPC201355-1G
6-Fluoro-1H-benzo[d]imidazole-2-carbaldehyde 95+%
6-Fluoro-1H-benzo[d]imidazole-2-carbaldehyde
Formula: C₈H₅FN₂O
MW: 164,137 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07778419
CAS Number: 885280-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR