Ammonium 2-aminohexafluoroisopropoxide 97%

Supplier: Apollo Scientific

PC2053-25G PC2053-5G
APOSPC2053-25GEA 306 EUR
APOSPC2053-25G APOSPC2053-5G
Ammonium 2-aminohexafluoroisopropoxide 97%
Ammonium 2-aminohexafluoroisopropoxide
Formula: C₃H₃F₆NO·H₃N
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD07784295
CAS Number: 1274903-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR