1,1,1-Trifluoro-2-(trifluoromethyl)pentan-2-ol
Supplier: Apollo Scientific
PC210043-1G
PC210043-500MG
APOSPC210043-1GEA
582
EUR
APOSPC210043-1G
APOSPC210043-500MG
1,1,1-Trifluoro-2-(trifluoromethyl)pentan-2-ol
1,1,1-Trifluoro-2-(trifluoromethyl)pentan-2-ol
Formula:
C₆H₈F₆O MW: 210.118 g/mol Boiling Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD28016154 CAS Number: 10315-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...