4-Bromo-2,5-difluorobenzaldehyde
Supplier: Apollo Scientific
|
Danger
|
PC210045-1G
PC210045-5G
PC210045-25G
APOSPC210045-1GEA
21.9
EUR
APOSPC210045-1G
APOSPC210045-5G
APOSPC210045-25G
4-Bromo-2,5-difluorobenzaldehyde
4-Bromo-2,5-difluorobenzaldehyde
Formula:
C₇H₃BrF₂O MW: 220.999 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847047 CAS Number: 357405-75-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...