Ethyl 4-amino-2-sulphanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate
Supplier: Apollo Scientific
PC210054-1G
APOSPC210054-1GEA
780
EUR
APOSPC210054-1G
Ethyl 4-amino-2-sulphanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate
Ethyl 4-amino-2-sulphanylidene-3-[3-(trifluoromethyl)phenyl]-2,3-dihydro-1,3-thiazole-5-carboxylate
Formula:
C₁₃H₁₁F₃N₂O₂S₂ MW: 348,364 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02089347 CAS Number: 312922-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...