1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}-1H-pyrazol-4-amine

Supplier: Apollo Scientific

PC210076-1G PC210076-500MG
APOSPC210076-1GEA 890 EUR
APOSPC210076-1G APOSPC210076-500MG
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}-1H-pyrazol-4-amine
1-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]methyl}-1H-pyrazol-4-amine
Formula: C₁₀H₈ClF₃N₄
Storage Temperature: Ambient
MDL Number: MFCD28347882
CAS Number: 1644602-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR