N-(4-Chlorophenyl)-2-{[1-methyl-4-(trifluoromethyl)-1H-pyrrol-3-yl]carbonyl}hydrazinecarbothioamide

Supplier: Apollo Scientific

PC210079-1G PC210079-500MG
APOSPC210079-1GEA 444 EUR
APOSPC210079-1G APOSPC210079-500MG
N-(4-Chlorophenyl)-2-{[1-methyl-4-(trifluoromethyl)-1H-pyrrol-3-yl]carbonyl}hydrazinecarbothioamide
N-(4-Chlorophenyl)-2-{[1-methyl-4-(trifluoromethyl)-1H-pyrrol-3-yl]carbonyl}hydrazinecarbothioamide
Formula: C₁₄H₁₂ClF₃N₄OS
MW: 376,784 g/mol
Melting Pt: 196…198 °C
Storage Temperature: Ambient
MDL Number: MFCD28369723
CAS Number: 1656294-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR