1,1,3,3-Tetrafluoroprop-1-ene

Supplier: Apollo Scientific
Danger

PC2258-1G PC2258-5G
APOSPC2258-1GEA 263 EUR
APOSPC2258-1G APOSPC2258-5G
1,1,3,3-Tetrafluoroprop-1-ene
1,1,3,3-Tetrafluoroprop-1-ene
Formula: C₃H₂F₄
MW: 114.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08457545
CAS Number: 4556-24-5
UN: 3161
ADR: 2.1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR