1-Bromo-2-fluoro-2-methylpropane 95%

Supplier: Apollo Scientific
Warning

PC2438-1G PC2438-500MG
APOSPC2438-1GEA 241 EUR
APOSPC2438-1G APOSPC2438-500MG
1-Bromo-2-fluoro-2-methylpropane 95%
1-Bromo-2-fluoro-2-methylpropane
Formula: C₄H₈BrF
MW: 155,009 g/mol
Boiling Pt: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD04972692
CAS Number: 19869-78-4
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR