2,3,4-Trifluorophenylhydrazine hydrochloride
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-Hydrazino-2,3,4-trifluorobenzene hydrochloride
PC2751-25G
PC2751-5G
PC2751-100G
APOSPC2751-25GEA
71.3
EUR
APOSPC2751-25G
APOSPC2751-5G
APOSPC2751-100G
2,3,4-Trifluorophenylhydrazine hydrochloride
2,3,4-Trifluorophenylhydrazine hydrochloride
Formula:
C₆H₅F₃N₂·ClH MW: 198,573 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094176 CAS Number: 502496-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...