2-Amino-4-bromo-6-fluorophenol 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-3-fluoro-2-hydroxyaniline

PC2775-1G PC2775-5G
APOSPC2775-1GEA 116 EUR
APOSPC2775-1G APOSPC2775-5G
2-Amino-4-bromo-6-fluorophenol 97%
2-Amino-4-bromo-6-fluorophenol
Formula: C₆H₅BrFNO
MW: 206.012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094184
CAS Number: 182499-89-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR