rac-Fmoc-Trifluoromethylalanine
Supplier: Apollo Scientific
PC27870-100MG
PC27870-1G
PC27870-250MG
APOSPC27870-100MGEA
713
EUR
APOSPC27870-100MG
APOSPC27870-1G
APOSPC27870-250MG
rac-Fmoc-Trifluoromethylalanine
rac-Fmoc-Trifluoromethylalanine
Formula:
C₁₈H₁₄F₃NO₄ MW: 365,303 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28898379 CAS Number: 1219349-78-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...