Fmoc-L-2,4,5-Trifluorophenylalanine
Supplier: Apollo Scientific
PC27885-100MG
PC27885-1G
PC27885-250MG
APOSPC27885-100MGEA
219
EUR
APOSPC27885-100MG
APOSPC27885-1G
APOSPC27885-250MG
Fmoc-L-2,4,5-Trifluorophenylalanine
Fmoc-L-2,4,5-Trifluorophenylalanine
Formula:
C₂₄H₁₈F₃NO₄ MW: 441.399 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659142 CAS Number: 959579-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...