5-Bromo-2,3-difluorobenzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC27917-250MG
APOSPC27917-250MGEA 79.9 EUR
APOSPC27917-250MG
5-Bromo-2,3-difluorobenzyl fluoride 95%
5-Bromo-2,3-difluorobenzyl fluoride
Formula: C₇H₄BrF₃
MW: 225,006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31664903
CAS Number: 2244085-63-8
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR