4-Bromo-2,5-difluorobenzyl fluoride 95%
Supplier: Apollo Scientific
|
Danger
|
PC27924-250MG
APOSPC27924-250MGEA
79.9
EUR
APOSPC27924-250MG
4-Bromo-2,5-difluorobenzyl fluoride 95%
4-Bromo-2,5-difluorobenzyl fluoride
Formula:
C₇H₄BrF₃ MW: 225,006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31664905 CAS Number: 2244083-92-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...