2-Chloro-5-fluoro-4-hydrazinylpyrimidine
Supplier: Apollo Scientific
|
Warning
|
PC28265-1G
PC28265-250MG
APOSPC28265-1GEA
261
EUR
APOSPC28265-1G
APOSPC28265-250MG
2-Chloro-5-fluoro-4-hydrazinylpyrimidine
2-Chloro-5-fluoro-4-hydrazinylpyrimidine
Formula:
C₄H₄ClFN₄ MW: 162,553 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16876982 CAS Number: 1314936-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...