2-(Ethylamino)-5-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific

Synonyms: N-[5-(Trifluoromethyl)pyridin-2-yl]ethylamine, N-Ethyl-5-(trifluoromethyl)pyridin-2-amine

PC300564-1G PC300564-250MG
APOSPC300564-1GEA 186 EUR
APOSPC300564-1G APOSPC300564-250MG
2-(Ethylamino)-5-(trifluoromethyl)pyridine 95%
2-(Ethylamino)-5-(trifluoromethyl)pyridine
Formula: C₈H₉F₃N₂
MW: 190,166 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD11118753
CAS Number: 89810-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR