1-(4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
Supplier: Apollo Scientific
PC300647-1G
PC300647-10G
PC300647-5G
APOSPC300647-1GEA
193
EUR
APOSPC300647-1G
APOSPC300647-10G
APOSPC300647-5G
1-(4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
1-(4-{[4-(Trifluoromethyl)pyrimidin-2-yl]oxy}phenyl)ethanone
Formula:
C₁₃H₉F₃N₂O₂ MW: 282,218 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11846260 CAS Number: 1227954-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...