4-Bromo-1-(4-fluorobenzyl)-1H-pyrazole

Supplier: Apollo Scientific
Warning

PC300652-1G PC300652-5G
APOSPC300652-1GEA 296 EUR
APOSPC300652-1G APOSPC300652-5G
4-Bromo-1-(4-fluorobenzyl)-1H-pyrazole
4-Bromo-1-(4-fluorobenzyl)-1H-pyrazole
Formula: C₁₀H₈BrFN₂
MW: 255,086 g/mol
Boiling Pt: 116 °C (2 mm Hg)
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD06167002
CAS Number: 251925-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR