2-(Aminomethyl)-4-(trifluoromethyl)pyrimidine ≥95%

Supplier: Apollo Scientific
Warning

Synonyms: N-Methyl-4-(trifluoromethyl)pyrimidin-2-amine

PC300670-1G PC300670-250MG
APOSPC300670-1GEA 290 EUR
APOSPC300670-1G APOSPC300670-250MG
2-(Aminomethyl)-4-(trifluoromethyl)pyrimidine ≥95%
2-(Aminomethyl)-4-(trifluoromethyl)pyrimidine
Formula: C₆H₆F₃N₃
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD13176794
CAS Number: 1193721-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR