4-Bromo-7-fluoro-1-benzofuran
Supplier: Apollo Scientific
|
Warning
|
PC300796-1G
PC300796-250MG
APOSPC300796-1GEA
219
EUR
APOSPC300796-1G
APOSPC300796-250MG
4-Bromo-7-fluoro-1-benzofuran
4-Bromo-7-fluoro-1-benzofuran
Formula:
C₈H₄BrFO MW: 215,019 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11877826 CAS Number: 1194376-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...