3-(3-Chloro-4-fluorophenyl)-1H-pyrazole
Supplier: Apollo Scientific
PC300891-1G
PC300891-250MG
APOSPC300891-1GEA
219
EUR
APOSPC300891-1G
APOSPC300891-250MG
3-(3-Chloro-4-fluorophenyl)-1H-pyrazole
3-(3-Chloro-4-fluorophenyl)-1H-pyrazole
Formula:
C₉H₆ClFN₂ Storage Temperature: Ambient |
MDL Number:
MFCD09860598 CAS Number: 154258-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...