3-Chloro-5-(trifluoromethyl)-1H-indazole

Supplier: Apollo Scientific
Warning

PC300921-1G PC300921-250MG
APOSPC300921-1GEA 621 EUR
APOSPC300921-1G APOSPC300921-250MG
3-Chloro-5-(trifluoromethyl)-1H-indazole
3-Chloro-5-(trifluoromethyl)-1H-indazole
Formula: C₈H₄ClF₃N₂
MW: 220,579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16995842
CAS Number: 1243407-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR