Methyl 5-fluoro-2,6-dimethoxynicotinate

Supplier: Apollo Scientific

PC300937-1G PC300937-5G
APOSPC300937-1GEA 339 EUR
APOSPC300937-1G APOSPC300937-5G
Methyl 5-fluoro-2,6-dimethoxynicotinate
Methyl 5-fluoro-2,6-dimethoxynicotinate
Formula: C₉H₁₀FNO₄
MW: 215,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529729
CAS Number: 2197053-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR