4-Fluoro-3-methylcinnamic acid 98%
Supplier: Apollo Scientific
PC302064-1G
PC302064-5G
APOSPC302064-1GEA
81.5
EUR
APOSPC302064-1G
APOSPC302064-5G
4-Fluoro-3-methylcinnamic acid 98%
4-Fluoro-3-methylcinnamic acid
Formula:
C₁₀H₉FO₂ MW: 180,176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03002834 CAS Number: 152152-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...