4-Fluoro-3-methyl-DL-phenylalanine

Supplier: Apollo Scientific
Warning

PC302151-1G PC302151-5G PC302151-10G PC302151-25G PC302151-100G
APOSPC302151-1GEA 128 EUR
APOSPC302151-1G APOSPC302151-5G APOSPC302151-10G APOSPC302151-25G APOSPC302151-100G
4-Fluoro-3-methyl-DL-phenylalanine
4-Fluoro-3-methyl-DL-phenylalanine
Formula: C₁₀H₁₂FNO₂
MW: 197,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116031
CAS Number: 1259956-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR