3,4,5-Trifluorobenzamidoxime 97%

Supplier: Apollo Scientific

PC302183-5G PC302183-1G
APOSPC302183-5GEA 94.5 EUR
APOSPC302183-5G APOSPC302183-1G
3,4,5-Trifluorobenzamidoxime 97%
3,4,5-Trifluorobenzamidoxime
Formula: C₇H₅F₃N₂O
MW: 190,123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07776441
CAS Number: 885954-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR