4-Chloro-3,5-difluorophenylacetonitrile

Supplier: Apollo Scientific
Danger

PC302326-1G PC302326-5G
APOSPC302326-1GEA 84.1 EUR
APOSPC302326-1G APOSPC302326-5G
4-Chloro-3,5-difluorophenylacetonitrile
4-Chloro-3,5-difluorophenylacetonitrile
Formula: C₈H₄ClF₂N
Storage Temperature: Ambient
MDL Number: MFCD09923910
CAS Number: 1000540-50-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR