2-Fluoro-5-methylbenzamide 98%
Supplier: Apollo Scientific
|
Warning
|
PC302766-5G
PC302766-1G
APOSPC302766-5GEA
192
EUR
APOSPC302766-5G
APOSPC302766-1G
2-Fluoro-5-methylbenzamide 98%
2-Fluoro-5-methylbenzamide
Formula:
C₈H₈FNO MW: 153,154 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04973775 CAS Number: 886500-04-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...