3-Chloro-5-fluoro-4-methoxybenzamide 97%

Supplier: Apollo Scientific

PC302772-10G PC302772-5G PC302772-1G
APOSPC302772-10GEA 386 EUR
APOSPC302772-10G APOSPC302772-5G APOSPC302772-1G
3-Chloro-5-fluoro-4-methoxybenzamide 97%
3-Chloro-5-fluoro-4-methoxybenzamide
Formula: C₈H₇ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD04115869
CAS Number: 886497-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR