3-Butoxy-6-chloro-2-fluorobenzaldehyde 97%

Supplier: Apollo Scientific

PC303054-1G PC303054-5G
APOSPC303054-1GEA 205 EUR
APOSPC303054-1G APOSPC303054-5G
3-Butoxy-6-chloro-2-fluorobenzaldehyde 97%
3-Butoxy-6-chloro-2-fluorobenzaldehyde
Formula: C₁₁H₁₂ClFO₂
MW: 230,663 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27986837
CAS Number: 1706430-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR