2,4-Dichloro-6-(pentafluorosulphur)aniline 97%
Supplier: Apollo Scientific
PC303151-1G
PC303151-5G
APOSPC303151-1GEA
350
EUR
APOSPC303151-1G
APOSPC303151-5G
2,4-Dichloro-6-(pentafluorosulphur)aniline 97%
2,4-Dichloro-6-(pentafluorosulphur)aniline
Formula:
C₆H₄Cl₂F₅NS MW: 288,066 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28054186 CAS Number: 1706446-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...