6-Ethoxy-2,3,4-trifluorobenzaldehyde 97%
Supplier: Apollo Scientific
PC303286-1G
PC303286-5G
APOSPC303286-1GEA
351
EUR
APOSPC303286-1G
APOSPC303286-5G
6-Ethoxy-2,3,4-trifluorobenzaldehyde 97%
6-Ethoxy-2,3,4-trifluorobenzaldehyde
Formula:
C₉H₇F₃O₂ MW: 204,146 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD29277472 CAS Number: 1980049-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...