4-Ethyl-2,3-difluorobenzaldehyde 97%

Supplier: Apollo Scientific
Warning

PC303297-5G PC303297-10G PC303297-1G
APOSPC303297-5GEA 447 EUR
APOSPC303297-5G APOSPC303297-10G APOSPC303297-1G
4-Ethyl-2,3-difluorobenzaldehyde 97%
4-Ethyl-2,3-difluorobenzaldehyde
Formula: C₉H₈F₂O
MW: 170,156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24686080
CAS Number: 1781592-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR