2-Chloro-4-fluoro-5-methylbenzyl alcohol 97%

Supplier: Apollo Scientific

PC303338-1G PC303338-5G
APOSPC303338-1GEA 290 EUR
APOSPC303338-1G APOSPC303338-5G
2-Chloro-4-fluoro-5-methylbenzyl alcohol 97%
2-Chloro-4-fluoro-5-methylbenzyl alcohol
Formula: C₈H₈ClFO
MW: 174,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24034286
CAS Number: 1536543-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR