6-Bromo-2,3-difluorobenzoyl chloride

Supplier: Apollo Scientific

PC303436-1G PC303436-5G
APOSPC303436-1GEA 203 EUR
APOSPC303436-1G APOSPC303436-5G
6-Bromo-2,3-difluorobenzoyl chloride
6-Bromo-2,3-difluorobenzoyl chloride
Formula: C₇H₂BrClF₂O
MW: 255,444 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18379809
CAS Number: 1263376-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR