5-Bromo-3-chloro-2-fluorophenylacetonitrile

Supplier: Apollo Scientific

PC303455-1G PC303455-5G
APOSPC303455-1GEA 325 EUR
APOSPC303455-1G APOSPC303455-5G
5-Bromo-3-chloro-2-fluorophenylacetonitrile
5-Bromo-3-chloro-2-fluorophenylacetonitrile
Formula: C₈H₄BrClFN
MW: 248.479 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26686147
CAS Number: 1537627-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR