5-Bromo-3-chloro-2-fluorophenylacetic acid

Supplier: Apollo Scientific

PC303456-1G PC303456-5G
APOSPC303456-1GEA 0 EUR
APOSPC303456-1G APOSPC303456-5G
5-Bromo-3-chloro-2-fluorophenylacetic acid
5-Bromo-3-chloro-2-fluorophenylacetic acid
Formula: C₈H₅BrClFO₂
MW: 267,48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26686407
CAS Number: 1537364-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR