5-Bromo-4-chloro-2-fluorophenylacetic acid
Supplier: Apollo Scientific
PC303480-1G
PC303480-5G
APOSPC303480-1GEA
799
EUR
APOSPC303480-1G
APOSPC303480-5G
5-Bromo-4-chloro-2-fluorophenylacetic acid
5-Bromo-4-chloro-2-fluorophenylacetic acid
Formula:
C₈H₅BrClFO₂ MW: 267,48 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640081 CAS Number: 1780668-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...