5-Bromo-4-chloro-2-fluorobenzylamine
Supplier: Apollo Scientific
PC303481-1G
PC303481-5G
APOSPC303481-1GEA
418
EUR
APOSPC303481-1G
APOSPC303481-5G
5-Bromo-4-chloro-2-fluorobenzylamine
5-Bromo-4-chloro-2-fluorobenzylamine
Formula:
C₇H₆BrClFN MW: 238,485 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28060767 CAS Number: 1780235-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...