3-Bromo-2,5,6-trifluorophenylacetonitrile

Supplier: Apollo Scientific

PC303509-1G PC303509-5G
APOSPC303509-1GEA 326 EUR
APOSPC303509-1G APOSPC303509-5G
3-Bromo-2,5,6-trifluorophenylacetonitrile
3-Bromo-2,5,6-trifluorophenylacetonitrile
Formula: C₈H₃BrF₃N
MW: 250,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640102
CAS Number: 1499733-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR