5-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 97%

Supplier: Apollo Scientific

PC303549-1G PC303549-5G
APOSPC303549-1GEA 371 EUR
APOSPC303549-1G APOSPC303549-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₂BrF₄N
MW: 268,006 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30829166
CAS Number: 2021202-81-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR