3-Bromo-2-chloro-5-(trifluoromethyl)benzylamine
Supplier: Apollo Scientific
PC303580-1G
PC303580-5G
APOSPC303580-1GEA
418
EUR
APOSPC303580-1G
APOSPC303580-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzylamine
3-Bromo-2-chloro-5-(trifluoromethyl)benzylamine
Formula:
C₈H₆BrClF₃N MW: 288.492 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32640157 CAS Number: 2384850-52-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...