5-Bromo-2-methyl-3-(trifluoromethyl)benzonitrile 97%

Supplier: Apollo Scientific

PC303595-1G PC303595-5G
APOSPC303595-1GEA 384 EUR
APOSPC303595-1G APOSPC303595-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzonitrile 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzonitrile
Formula: C₉H₅BrF₃N
MW: 264,042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32640169
CAS Number: 2387259-19-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR