4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
N,N-Dimethyl-2-(trifluoroacetyl)ethenylamine, N,N-Dimethyl-2-(trifluoroacetyl)vinylamine
PC31140-1G
PC31140-5G
APOSPC31140-1GEA
163
EUR
APOSPC31140-1G
APOSPC31140-5G
4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
4-(Dimethylamino)-1,1,1-trifluorobut-3-en-2-one
Formula:
C₆H₈F₃NO MW: 167.129 g/mol Melting Pt: 56…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00764476 CAS Number: 142503-25-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...