Ethyl 2-bromo-2-(4-fluorophenyl)acetate
Supplier: Apollo Scientific
|
Warning
|
PC31351-1G
PC31351-250MG
APOSPC31351-1GEA
39.3
EUR
APOSPC31351-1G
APOSPC31351-250MG
Ethyl 2-bromo-2-(4-fluorophenyl)acetate
Ethyl 2-bromo-2-(4-fluorophenyl)acetate
Formula:
C₁₀H₁₀BrFO₂ MW: 261,088 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00204082 CAS Number: 712-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...