1-Methyl-3-(trifluoroacetyl)-1H-indole

Supplier: Apollo Scientific
Warning

Synonyms: 2,2,2-Trifluoro-1-(1-methyl-1H-indol-3-yl)ethan-1-one

PC32714-1G PC32714-5G
APOSPC32714-1GEA 69.6 EUR
APOSPC32714-1G APOSPC32714-5G
1-Methyl-3-(trifluoroacetyl)-1H-indole
1-Methyl-3-(trifluoroacetyl)-1H-indole
Formula: C₁₁H₈F₃NO
MW: 227.183 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02571216
CAS Number: 318-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR