2-Bromo-5-fluorophenacyl bromide 94+%

Supplier: Apollo Scientific
Danger

Synonyms: 2,2'-Dibromo-5'-fluoroacetophenone, 2-Bromo-1-(2-bromo-5-fluorophenyl)ethan-1-one

PC32728-1G PC32728-250MG
APOSPC32728-1GEA 466 EUR
APOSPC32728-1G APOSPC32728-250MG
2-Bromo-5-fluorophenacyl bromide 94+%
2-Bromo-5-fluorophenacyl bromide
Formula: C₈H₅Br₂FO
MW: 295,931 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23711741
CAS Number: 1427446-94-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR