5-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
PC32789-1G
PC32789-250MG
APOSPC32789-1GEA
118
EUR
APOSPC32789-1G
APOSPC32789-250MG
5-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
5-Fluoro-2-oxo-2H-chromene-3-carboxylic acid
Formula:
C₁₀H₅FO₄ MW: 208,143 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26684685 CAS Number: 1540476-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...