tert-Butyl (3-fluoropropyl)carbamate 95%
Supplier: Apollo Scientific
|
Warning
|
PC32843-1G
PC32843-250MG
APOSPC32843-1GEA
786
EUR
APOSPC32843-1G
APOSPC32843-250MG
tert-Butyl (3-fluoropropyl)carbamate 95%
tert-Butyl (3-fluoropropyl)carbamate
Formula:
C₈H₁₆FNO₂ MW: 177,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23703459 CAS Number: 178181-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...